Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AKR7A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584658
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584658 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584658 Supplier Invitrogen™ Supplier No. PA584658
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LHQEGKFVEL GLSNYAAWEV AEICTLCKSN GWILPTVYQ Highest antigen sequence indentity to the following orthologs: Rat - 95%, Mouse - 97%.

AKR7A2 is aldo-keto reductases, which are involved in the detoxification of aldehydes and ketones.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AKR7A2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AKR7A2
Gene Accession No. O43488
Gene Alias 0610025K21Rik; Afar; AFAR1; Afar2; AFB1 aldehyde reductase 1; AFB1-AR 1; AFB1-AR1; aflatoxin aldehyde reductase; aflatoxin B1 aldehyde reductase member 2; aflatoxin beta1 aldehyde reductase; Aiar; AKR7; AKR7A2; Akr7a5; aldo-keto reductase family 7 member A2; aldo-keto reductase family 7, member A2 (aflatoxin aldehyde reductase); aldo-keto reductase family 7, member A5 (aflatoxin aldehyde reductase); aldoketoreductase 7; epididymis secretory sperm binding protein Li 166mP; HEL-S-166mP; rAFAR2; SSA reductase; succinic semialdehyde reductase; synonyms: AFAR1
Gene Symbols AKR7A2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human AKR7A2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8574
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.