Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AKR7A2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | AKR7A2 |
---|---|
Concentration | 0.2 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-6547
![]() ![]() |
Proteintech Group Inc
107301AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
AKR7A2 is aldo-keto reductases, which are involved in the detoxification of aldehydes and ketones.Specifications
AKR7A2 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
O43488, Q8CG76 | |
110198, 8574 | |
AKR7A2 Fusion Protein Ag1049 | |
Primary | |
-20°C | |
AKR7A2 |
0.2 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
AFAR, AFAR1, AFB1 aldehyde reductase 1, AFB1 AR 1, AFB1 AR1, AKR7, AKR7A2, Aldoketoreductase 7, SSA reductase | |
AKR7A2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title