Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AKR7A3 Antibody (2E11), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197944AF532
Description
AKR7A3 Monoclonal specifically detects AKR7A3 in Human samples. It is validated for Western Blot, ELISA.Specifications
AKR7A3 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
AKR7A3 | |
Recombinant human AKR7A3 (1-331aa) purified from E. coli | |
0.1 mL | |
22977.0 | |
Store at 4°C in the dark. |
Western Blot, ELISA | |
200000000000 | |
Western Blot, ELISA | |
AFAR2, AFB1 aldehyde reductase 2, AFB1-AR 2, aflatoxin B1 aldehyde reductase 2, aflatoxin B1 aldehyde reductase member 3, aldo-keto reductase family 7, member A3 (aflatoxin aldehyde reductase) | |
Mouse | |
Protein G purified | |
Primary | |
Human | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction