Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AKR7A3 Antibody (2E11), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197944AF700
Description
AKR7A3 Monoclonal specifically detects AKR7A3 in Human samples. It is validated for Western Blot, ELISA.Specifications
| AKR7A3 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| AKR7A3 | |
| Recombinant human AKR7A3 (1-331aa) purified from E. coli | |
| 100 μL | |
| Primary | |
| Anti-human AKR7A3 mAb, clone 2E11, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human AKR7A3 protein. | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA | |
| 200000000000 | |
| Western Blot, ELISA | |
| AFAR2, AFB1 aldehyde reductase 2, AFB1-AR 2, aflatoxin B1 aldehyde reductase 2, aflatoxin B1 aldehyde reductase member 3, aldo-keto reductase family 7, member A3 (aflatoxin aldehyde reductase) | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 22977 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction