Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AKR7A3 Antibody (2E11), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197944C
Description
AKR7A3 Monoclonal specifically detects AKR7A3 in Human samples. It is validated for Western Blot, ELISA.Specifications
AKR7A3 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
AKR7A3 | |
Recombinant human AKR7A3 (1-331aa) purified from E. coli | |
100 μL | |
Primary | |
Anti-human AKR7A3 mAb, clone 2E11, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human AKR7A3 protein. | |
Store at 4C in the dark. | |
IgG1 κ |
Western Blot, ELISA | |
200000000000 | |
Western Blot, ELISA | |
AFAR2, AFB1 aldehyde reductase 2, AFB1-AR 2, aflatoxin B1 aldehyde reductase 2, aflatoxin B1 aldehyde reductase member 3, aldo-keto reductase family 7, member A3 (aflatoxin aldehyde reductase) | |
Mouse | |
Protein G purified | |
RUO | |
22977 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction