Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AKR7A3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528482
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528482 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528482 Supplier Invitrogen™ Supplier No. PA528482
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, HeLaS3, HepG2, Molt-4, Raji, Mammary gland tumor tissues. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Aldo-keto reductases, such as AKR7A3, are involved in the detoxification of aldehydes and ketones.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AKR7A3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene AKR7A3
Gene Accession No. O95154
Gene Alias Afar; AFAR2; AFB1 aldehyde reductase 2; AFB1-AR; AFB1-AR 2; aflatoxin aldehyde reductase; aflatoxin B1 aldehyde reductase 2; aflatoxin B1 aldehyde reductase member 1; Aflatoxin B1 aldehyde reductase member 3; Akr7a1; AKR7A3; aldo-keto reductase family 7 member A3; aldo-keto reductase family 7, member A3 (aflatoxin aldehyde reductase); rAFAR1
Gene Symbols AKR7A3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human AKR7A3. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22977
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.