Learn More
Invitrogen™ AKR7A3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596969
Description
Immunogen sequence: MSRQLSRARP ATVLGAMEMG RRMDAPTSAA VTRAFLERGH TEIDTAFVYS EGQSETILGG LGLRLGGSDC RVKIDTKAIP LFGNSLKPDS LRFQLETSLK RLQCPRVDLF YLHMPDHSTP VEETLRACHQ LHQEGKFVEL GLSNYAAWEV AEICTLCKSN GWILPTVYQG MYNAITRQVE TELFPCLRHF GLRFYAFNPL AGGLLTGKYK YEDKNGKQPV GRFFGNTWAE MYRNRYWKEH HFEGIALVEK ALQAAYGASA PSMTSATLRW MYHHSQLQGA HGDAVILGMS SLEQLEQNLA AAEEGPLEPA VVDAFNQAWH LVTHECPNYF R; Positive Samples: HepG2, HT-29, Mouse kidney, Mouse liver, Mouse pancreas; Cellular Location: Cytoplasm.
Aldo-keto reductases, such as AKR7A3, are involved in the detoxification of aldehydes and ketones.
Specifications
AKR7A3 | |
Polyclonal | |
Unconjugated | |
AKR7A3 | |
Afar; AFAR2; AFB1 aldehyde reductase 2; AFB1-AR; AFB1-AR 2; aflatoxin aldehyde reductase; aflatoxin B1 aldehyde reductase 2; aflatoxin B1 aldehyde reductase member 1; Aflatoxin B1 aldehyde reductase member 3; Akr7a1; AKR7A3; aldo-keto reductase family 7 member A3; aldo-keto reductase family 7, member A3 (aflatoxin aldehyde reductase); rAFAR1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
22977, 26760 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O95154, P38918 | |
AKR7A3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-331 of human AKR7A3 (NP_0361992). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.