Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AKR7L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572255
Description
AKR7L Polyclonal Antibody for Western Blot, ICC/IF
Can reduce the dialdehyde protein-binding form of aflatoxin B1 (AFB1) to the non-binding AFB1 dialcohol. May be involved in protection of liver against the toxic and carcinogenic effects of AFB1, a potent hepatocarcinogen.
Specifications
AKR7L | |
Polyclonal | |
Unconjugated | |
AKR7L | |
AFAR3; AFB1 aldehyde reductase 3; AFB1-AR 3; AFB1-AR3; Aflatoxin B1 aldehyde reductase member 4; AKR7A4; AKR7L; aldo-keto reductase family 7 like (gene/pseudogene); Aldoketoreductase 7-like | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
246181 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.44 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q8NHP1 | |
AKR7L | |
KLH conjugated synthetic peptide between 32-64 amino acids from human AKR7L. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction