Learn More
Description
AKT Inhibitor IV is weakly fluorescent upon irradiation with UV light (excitation/emission maxima λ ∼ 388/460; quantum yield = 0.0016).
Specifications
Specifications
| Chemical Name or Material | 6-(2-Benzothiazolyl)-1-ethyl-2-[(1 E)-2-(methylphenylamino)ethenyl]-3-phenyl-1 H-benzimidazolium iodide |
| CAS | 959841-49-7 |
| Quantity | 5 mg |
| Product Line | Tocris |
| Target | Akt (Protein Kinase B) Inhibitors |
| Molecular Formula | C31H27IN4S |
| Purity | 0.98 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

