Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKT Monoclonal Antibody (14E5.16C8.25F6), Rockland™
SDP

Catalog No. 501990532
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0532 25 μL
50-199-0531 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-0532 Supplier Rockland Immunochemicals Supplier No. 200301401S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This antibody is specific for human and mouse AKT protein. A BLAST analysis was used to suggest cross-reactivity with AKT1, AKT2, and AKT3 from human, mouse, rat and chimpanzee sources based on 100% homology with the immunizing sequence. Cross-reactivity with AKT1, 2, 3 was determined with Western Blot. Cross reactivity of AKT from other sources has not been determined.

AKT also known as protein kinase B (PKB) or RAS-alpha, is an ubiquitous serine/threonine kinase that plays an important role in diverse biological responses such as regulation of metabolism, cell survival and growth by phosphorylating multiple proteins. This protein kinase is activated by insulin, PI3K, IGF1 and various other growth and survival factors. Akt promotes cell survival by inhibiting apoptosis through phosphorylation and inactivation of several targets, including forkhead transcription factors, and caspase-9. The AKT pathway is a major target for cancer drug discovery.

Specifications

Antigen AKT
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 14E5.16C8.25F6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene AKT1
Gene Accession No. P31749, P31750, P31751, P47196, P47197, Q60823, Q63484, Q9WUA6, Q9Y243
Gene Alias 2410016A19Rik; AI851531; Akt; Akt kinase; AKT serine/threonine kinase 1; AKT serine/threonine kinase 2; AKT serine/threonine kinase 3; AKT/PKB; Akt1; AKT1 kinase; AKT1m; Akt1m protein; Akt1-PA; Akt1-PB; Akt1-PC; Akt1-PD; Akt1-PE; AKT2; AKT2 kinase; Akt3; AKT3 kinase; akt3a; AW554154; cb945; CG4006; CG4006 gene product from transcript CG4006-RE; CG4006-PA; CG4006-PB; CG4006-PC; CG4006-PD; CG4006-PE; CWS6; D930002M15Rik; DAkt; D-Akt; dAkt kinase; dAKT/dPKB; dAkt/PKB; dAkt1; DAKT1/PKB; DKFZp434N0250; Dmel\CG4006; Dmel_CG4006; Dpkb; DRAC-PK; DRAC-PK66; DRAC-PK85; HIHGHH; kinase Akt1; l(3)04226; l(3)89Bq; MGC99656; MPPH; MPPH2; murine thymoma viral (v-akt) homolog-2; murine thymoma viral (v-akt) oncogene homolog 1; murine thymoma viral (v-akt) oncogene homolog 2; Nmf350; pAkt; p-Akt; PKB; PKB alpha; PKB beta; PKB gamma; PKB/Akt; PKB/dAKT; PKBalpha; PKB-ALPHA; PKBB; PKBbeta; PKBG; PKB-GAMMA; PRKBA; PRKBB; PRKBG; Protein kinase Akt-2; Protein kinase Akt-3; Protein kinase B; Protein kinase B a
Gene Symbols AKT1, Akt2, Akt3
Host Species Mouse
Immunogen Anti-AKT Antibody was produced by repeated immunizations in mice with a synthetic peptide corresponding to residues internal to the human AKT1 protein.
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10000, 100609583, 11651, 11652, 207, 208, 23797, 24185, 25233, 29414, 456035, 740898
Target Species Chimpanzee, Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.