Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat Akt Pan Specific Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC2055R100UG
This item is not returnable.
View return policy
Description
AKT Monoclonal specifically detects AKT in Human, Mouse, Rat samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
AKT | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
P31749 | |
AKT1 | |
E. coli-derived recombinant human Akt1 Ser2-Ala480 Accession # P31749 | |
RUO | |
207 | |
Human, Mouse, Rat | |
IgG2b |
Flow Cytometry | |
281046 | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
RAC-ALPHA, AKT, AKT serine/threonine kinase 1, PKB, PKB alpha, PKB beta, PKB gamma, PKB-ALPHA, PRKBA, RAC, RAC-alpha, RAC-beta, RAC-gamma | |
Mouse | |
100 μg | |
Primary | |
Detects human, mouse and rat Akt in direct ELISAs and Western blots. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction