Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Akt/PKB, PH Domain, Mouse, Alexa Fluor 647, Clone: SKB1,

Catalog No. 05591AF647 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Alexa Fluor 647
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
05591AF647 100 μL Alexa Fluor 647
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05591AF647 Supplier MilliporeSigma™ Supplier No. 05591AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RAC-alpha serine/threonine-protein kinase (EC 2.7.11.1; UniProt P31749; also known as PKB, PKB alpha, Protein kinase B, Protein kinase B alpha, Proto-oncogene c-Akt, RAC-PK-alpha) is encoded by the AKT1 (also known as PKB, RAC) gene (Gene ID 207) in human. AKTs (protein kinase Bs) are serine/threonine kinases that play an important role in signal transduction pathways involved in cell proliferation, apoptosis, angiogenesis, and diabetes. There exit three types of AKTs (α, β, γ or AKT 1, 2, 3) sharing a high degree of homology, but differing slightly in the localization of their regulatory phosphorylation sites. AKT α is the predominant isoform in most tissues, whereas the highest expression of AKT β is observed in the insulin-responsive tissues, and AKT γ is abundant in brain tissue. Each AKT type is composed of three functionally distinct regions, including an N-terminal pleckstrin homology (PH) domain that provides a lipid-binding module to direct Akt to PIP2 and PIP3, a central catalytic domain, and a C-terminal hydrophobic motif.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Akt/PKB, PH Domain
Applications Immunocytochemistry
Classification Monoclonal
Clone SKB1
Concentration Please refer to lot specific datasheet.
Conjugate Alexa Fluor 647
Formulation Purified mouse monoclonal IgG antibody conjugate in PBS with 15 mg/mL BSA and 0.1 % Sodium Azide.
Gene AKT1, PKB, RAC
Gene Accession No. P31749
Host Species Mouse
Immunogen GST-fusion protein corresponding to residues 1-149 of human Akt1.
Purification Method Protein G Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Apoptosis & Cancer
Primary or Secondary Primary
Gene ID (Entrez) NM_005163
Test Specificity Clone SKB1 recognizes Akt1 and Akt2, but not Akt3, GST fusion proteins at 100 ng by immunoblotting analysis using the unconjugated antibody (Mfr. No. 05-591).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.