Learn More
AKT Polyclonal Antibody, Rockland™

Sheep Polyclonal Antibody
$174.73 - $636.24
Specifications
Antigen | AKT |
---|---|
Concentration | 0.9 mg/mL |
Applications | ELISA, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-2569
![]() ![]() |
Rockland Immunochemicals
600601401 |
100 μg |
Each for $636.24
|
|
|||||
50-199-2570
![]() ![]() |
Rockland Immunochemicals
600601401S |
25 μL |
Each for $174.73
|
|
|||||
Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This antibody is directed against AKT and reacts with the AKT from human tissues. Based on sequence we expect this antibody to react as well with rat, mouse and chicken AKT.
AKT also known as protein kinase B (PKB) or RAS-alpha, is an ubiquitous serine/threonine kinase that plays an important role in diverse biological responses such as regulation of metabolism, cell survival and growth by phosphorylating multiple proteins. This protein kinase is activated by insulin, PI3K, IGF1 and various other growth and survival factors. Akt promotes cell survival by inhibiting apoptosis through phosphorylation and inactivation of several targets, including forkhead transcription factors, and caspase-9. The AKT pathway is a major target for cancer drug discovery.Specifications
AKT | |
ELISA, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
Sheep | |
Human | |
P31749, P31751, Q9Y243 | |
10000, 207, 208 | |
Primary | |
Antibody |
0.9 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
0.02M potassium phosphate with 0.15M NaCl and 0.1% sodium azide; pH 7.2 | |
2410016A19Rik; AI851531; Akt; Akt kinase; AKT serine/threonine kinase 1; AKT serine/threonine kinase 2; AKT serine/threonine kinase 3; AKT/PKB; Akt1; AKT1 kinase; AKT1m; Akt1m protein; Akt1-PA; Akt1-PB; Akt1-PC; Akt1-PD; Akt1-PE; AKT2; AKT2 kinase; Akt3; AKT3 kinase; akt3a; AW554154; cb945; CG4006; CG4006 gene product from transcript CG4006-RE; CG4006-PA; CG4006-PB; CG4006-PC; CG4006-PD; CG4006-PE; CWS6; D930002M15Rik; DAkt; D-Akt; dAkt kinase; dAKT/dPKB; dAkt/PKB; dAkt1; DAKT1/PKB; DKFZp434N0250; Dmel\CG4006; Dmel_CG4006; Dpkb; DRAC-PK; DRAC-PK66; DRAC-PK85; HIHGHH; kinase Akt1; l(3)04226; l(3)89Bq; MGC99656; MPPH; MPPH2; murine thymoma viral (v-akt) homolog-2; murine thymoma viral (v-akt) oncogene homolog 1; murine thymoma viral (v-akt) oncogene homolog 2; Nmf350; pAkt; p-Akt; PKB; PKB alpha; PKB beta; PKB gamma; PKB/Akt; PKB/dAKT; PKBalpha; PKB-ALPHA; PKBB; PKBbeta; PKBG; PKB-GAMMA; PRKBA; PRKBB; PRKBG; Protein kinase Akt-2; Protein kinase Akt-3; Protein kinase B; Protein kinase B a | |
AKT1, Akt2, Akt3 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Akt2 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.