Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Akt (pS473) Mouse anti-Human, Mouse, R718, Clone: M89-61, BD Phosflow™

Catalog No. BDB567634 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB567634 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BDB567634 Supplier BD Biosciences Supplier No. 567634
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Akt (pS473) Monoclonal antibody specifically detects Akt (pS473) in Human, Mouse samples. It is validated for Intracellular staining (Flow Cytometry)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Akt (pS473)
Applications Flow Cytometry
Classification Monoclonal
Clone M89-61
Conjugate R718
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Akt1, Akt2, Akt3, PKBα, PKBβ, PKBγ, RAC-PKα, RAC-PKβ, RAC-PKγ, STK-2
Host Species Mouse
Immunogen Phosphorylated Human Akt1 (pS473) Peptide
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.