Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AKT Rabbit anti-Human, Mouse, Rat, Clone: 2O2W6, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $572.50
Specifications
Antigen | AKT |
---|---|
Clone | 2O2W6 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
AKT Monoclonal antibody specifically detects AKT in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
AKT | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cellular Signaling, Neuroscience | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
207 | |
IgG | |
Affinity purified |
2O2W6 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
RAC-ALPHA, AKT, AKT serine/threonine kinase 1, PKB, PKB-ALPHA, PRKBA, RAC | |
A synthetic phosphorylated peptide around T308 of human AKTAKT1 (NP_005154.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title