Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ AKT1/2/3 Inhibitor Peptide Set
SDP

Catalog No. NBP2293325 Shop All Novus Products
Change view
Click to view available options
Quantity:
2 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2293325 5 mg
NBP229332 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP2293325 Supplier R&D Systems™ Supplier No. NBP2293325MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. For use in research applications

Specifications

Host Species Human
Components Akt (Isoforms 1,2,3) Inhibitor peptide: 2 x 1mg (lyophilized) DRQIKIWFQNRRMKWKKAVTDHPDRLWAWEKF (AKT sequence: AVTDHPDRLWAWEKF). Molecular weight: 4214. Antennapedia Control peptide: 2 x 1mg (lyophilized) DRQIKIWFQNRRMKWKK. Molecular weight: 2361
For Use With (Application) Inhibition of Akt kinase activity
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Quantity 5 mg
Product Type AKT1/2/3 Inhibitor Peptide Set
Molecular Weight (g/mol) 4214
Product Line Novus
Inhibitors AKT1/2/3
Form Lyophilized

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.