Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AKT1/2/3 Rabbit anti-Human, Mouse, Rat, Clone: 2E3A3, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $550.00
Specifications
Antigen | AKT1/2/3 |
---|---|
Clone | 2E3A3 |
Dilution | Western Blot 1:500 - 1:1000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
AKT1/2/3 Monoclonal antibody specifically detects AKT1/2/3 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
AKT1/2/3 | |
Western Blot 1:500 - 1:1000 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
AKT, EC 2.7.11, EC 2.7.11.1, PKBMGC99656, PRKBA, Protein kinase B, Proto-oncogene c-Akt, rac protein kinase alpha, RAC-ALPHA, RAC-alpha serine/threonine-protein kinase, RAC-PK-alpha, RACPKB-ALPHA, v-akt murine thymoma viral oncogene homolog 1 | |
A synthetic phosphorylated peptide around S473 of human AKT1/2/3 (NP_005154.2). QFSYS | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
2E3A3 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Apoptosis, Autophagy, Cancer, Growth and Development, Hypoxia, mTOR Pathway, Neuronal Cell Markers, Phospho Specific, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
207 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title