Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AKT1 Antibody (V7), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250101C
Description
AKT1 Monoclonal specifically detects AKT1 in Zebrafish samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
AKT1 | |
Monoclonal | |
DyLight 650 | |
50 mM Sodium Borate with 0.05% Sodium Azide | |
AKT1 | |
This AKT1 antibody was made to an ovalbumin-conjugated synthetic peptide KDPMQRLGGG (C-terminal sequence) | |
Protein A purified | |
RUO | |
Primary | |
Zebrafish | |
Purified |
Western Blot, ELISA, Immunohistochemistry | |
V7 | |
Western Blot, ELISA, Immunohistochemistry | |
AKT, AKT serine/threonine kinase 1, AKT1m, CWS6, EC 2.7.11, EC 2.7.11.1, PKB, PKB alpha, PKBMGC99656, PRKBA, Protein kinase B, Protein Kinase B Alpha, Proto-oncogene c-Akt, RAC, rac protein kinase alpha, RAC-ALPHA, RAC-alpha serine/threonine-protein kinas | |
Mouse | |
55.7 kDa | |
0.1 mL | |
Apoptosis, Autophagy, Cancer, Growth and Development, Hypoxia, mTOR Pathway, Neuronal Cell Markers, Signal Transduction | |
207 | |
Store at 4C in the dark. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction