Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AKT1 Monoclonal Antibody (OTI3E3), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA504231

Description
The Akt family of serine/threonine protein kinases (also called protein kinase B/PKB) plays a key role in insulin signaling, cellular survival, and transformation. Akt1 is activated by phosphorylation on residues threonine 308 and Serine 473. Thr308, in the catalytic domain, is phosphorylated by the protein kinase PDK1, and Ser473, in the COOH-terminal regulatory domain, is phosphorylated by putative serine 473 kinase. To date, a couple of proteins have been shown to physically interact with PKB.Specifications
AKT1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P31749, P31750, P47196 | |
AKT1 | |
Full length human recombit protein of human AKT1 produced in HEK293T cell. | |
100 μL | |
Primary | |
Canine, Human, Mouse, Rat | |
Antibody | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI3E3 | |
Unconjugated | |
AKT1 | |
AKT, CWS6, PKB, PKB-ALPHA, PRKBA, RAC, RAC-ALPHA | |
Mouse | |
Affinity Chromatography | |
RUO | |
11651, 207, 24185, 490878 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction