Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKT1 Mouse anti-Human, Clone: OTI2B9, lyophilized, TrueMAB™
SDP

Catalog No. 501662735
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-2735 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-2735 Supplier OriGene Supplier No. CF504236
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

The Akt family of serine/threonine protein kinases (also called protein kinase B/PKB) plays a key role in insulin signaling, cellular survival, and transformation. Akt1 is activated by phosphorylation on residues threonine 308 and Serine 473. Thr308, in the catalytic domain, is phosphorylated by the protein kinase PDK1, and Ser473, in the COOH-terminal regulatory domain, is phosphorylated by putative serine 473 kinase. To date, a couple of proteins have been shown to physically interact with PKB.

Specifications

Antigen AKT1
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone OTI2B9
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene AKT1
Gene Accession No. P31749
Gene Alias AKT, CWS6, PKB, PKB-ALPHA, PRKBA, RAC, RAC-ALPHA
Gene Symbols AKT1
Host Species Mouse
Immunogen Full length human recombit protein of human AKT1 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 207
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.