Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKT1 Mouse anti-Human, Mouse, Rat, APC, Clone: 5E5.F5.D7, Rockland™
SDP

Catalog No. 501990726
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0726 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-0726 Supplier Rockland Immunochemicals Supplier No. 200326J34
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Store vial at 4°C prior to restoration. Restore with deionized water (or equivalent). This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Centrifuge product if not completely clear after standing at room temperature. For extended storage, aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Store reagent in the dark. Use subdued lighting during handling and incubation of cells prior to analysis. Anti-AKT1 antibody is directed against human AKT1. The antibody detects both unphosphorylated and phosphorylated forms of the protein. Cross reactivity with AKT1 from other species has not been determined, however, the sequence of the immunogen shows 85% identity to mouse and 92% identity with rat, therefore, cross reactivity is expected.

AKT also known as protein kinase B (PKB) or RAS-alpha, is an ubiquitous serine/threonine kinase that plays an important role in diverse biological responses such as regulation of metabolism, cell survival and growth by phosphorylating multiple proteins. This protein kinase is activated by insulin, PI3K, IGF1 and various other growth and survival factors. Akt promotes cell survival by inhibiting apoptosis through phosphorylation and inactivation of several targets, including forkhead transcription factors, and caspase-9. The AKT pathway is a major target for cancer drug discovery.

Specifications

Antigen AKT1
Applications Dot Blot, ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone 5E5.F5.D7
Concentration 1 mg/mL
Conjugate APC
Formulation 0.02M potassium phosphate with 0.5M NaCl, 10 mg/mL BSA and no preservative; pH 7.2
Gene AKT1
Gene Accession No. P31749, P31750, P47196
Gene Alias Akt; Akt kinase; AKT serine/threonine kinase 1; AKT/PKB; AKT1; AKT1 kinase; AKT1m; Akt1m protein; Akt1-PA; Akt1-PB; Akt1-PC; Akt1-PD; Akt1-PE; AKT2; AKT3; CAKT; C-AKT; CG4006; CG4006 gene product from transcript CG4006-RE; CG4006-PA; CG4006-PB; CG4006-PC; CG4006-PD; CG4006-PE; CWS6; DAkt; D-Akt; dAkt kinase; dAKT/dPKB; dAkt/PKB; DAkt1; DAKT1/PKB; Dmel\CG4006; Dmel_CG4006; dPKB; DRAC-PK; DRAC-PK66; DRAC-PK85; kinase Akt1; l(3)04226; l(3)89Bq; MGC99656; murine thymoma viral (v-akt) oncogene homolog 1; pAkt; p-Akt; Phospho-AKT; Phospho-PKB; PKB; PKB Akt; PKB alpha; PKB beta; PKB gamma; PKB/AKT; PKB/dAKT; PKBalpha; PKB-ALPHA; PKBG; PRKBA; Protein kinase B; Protein kinase B alpha; protein kinase B-alpha; Proto-oncogene c-Akt; Rac; rac protein kinase alpha; RAC protein kinase alpha RAC-PK alpha; RAC serine/threonine-protein kinase; RAC-ALPHA; RAC-alpha serine/threonine-protein kinase; RacPK; RAC-PK-alpha; RAC-PK-beta; RAC-PK-gamma; related to A and C kinases; related to PKA to PKC protein kinases; related to the A and C kinases; serine/threonine protein kinase; serine-threonine protein kinase; STK-2; Thymoma viral proto-oncogene; thymoma viral proto-oncogene 1; v-akt murine thymoma viral oncogene homolog 1; v-akt murine thymoma viral oncogene-like protein 1
Gene Symbols AKT1
Host Species Mouse
Immunogen Anti-AKT1 Antibody was produced in mice by repeated immunizations with a synthetic peptide corresponding to internal residues of human AKT1 protein followed by monoclonal development.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11651, 207, 24185
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.