Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKT1 Mouse anti-Human, Unconjugated, Clone: AKT1/2491, Abnova™

Catalog No. 89337436 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-436 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-436 Supplier Abnova Corporation Supplier No. MAB20941
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

AKT1 Monoclonal antibody specifically detects AKT1 in Human samples. It is validated for IHC-P.

Specifications

Antigen AKT1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone AKT1/2491
Conjugate Unconjugated
Description v-akt murine thymoma viral oncogene homolog 1
Gene AKT1
Gene Alias AKT, MGC99656, PKB, PKB-ALPHA, PRKBA, RAC, RAC-ALPHA
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 85-189 of human AKT1
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 207
Target Species Human
Content And Storage Store at 2 to 8°C
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.