Learn More
Description
This product is intended for use as a positive control in Western blot. You will receive the lysate (100 ug), and an empty vector negative control (100 ug).
Specifications
Specifications
| Cell Line | This AKT1 lysate is made in HEK293T cells, using plasmid ID RC201850 and based on accession number NM_001014431. The protein contains a C-terminal DDK tag. |
| Quantity | 0.1 mg |
| Storage Buffer | RIPA buffer |
| Purity | Protein |
| Test Specificity | Homo sapiens v-akt murine thymoma viral oncogene homolog 1 (AKT1), transcript variant 3, mRNA. |
| Content And Storage | -80°C. Avoid freeze-thaw cycles. |
| Product Line | Novus |
| Protein State | Native |
| For Use With (Application) | Western Blot |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
