Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AKT1, p Ser124 Antibody (SR2049), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | AKT1 |
---|---|
Clone | SR2049 |
Dilution | Western Blot 1:500-1:1000, Immunoprecipitation 1:50 |
Applications | Western Blot, Immunoprecipitation |
Classification | Monoclonal |
Description
AKT1 Monoclonal antibody specifically detects AKT1 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
AKT1 | |
Western Blot 1:500-1:1000, Immunoprecipitation 1:50 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
AKT, AKT serine/threonine kinase 1, AKT1m, CWS6, EC 2.7.11, EC 2.7.11.1, PKB, PKB alpha, PKBMGC99656, PRKBA, Protein kinase B, Protein Kinase B Alpha, Proto-oncogene c-Akt, RAC, rac protein kinase alpha, RAC-ALPHA, RAC-alpha serine/threonine-protein kinase, RAC-PK-alpha, RACPKB-ALPHA, Serine-Threonine Protein Kinase, v-akt murine thymoma viral oncogene homolog 1, V-Akt Murine Thymoma Viral Oncogene-Like Protein 1 | |
A synthesized peptide derived from human phospho-AKT1 [Ser124] (Uniprot #: P31749) | |
Primary | |
Store at -20 degrees C. Avoid freeze/thaw cycles. |
SR2049 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Angiogenesis, Apoptosis, Autophagy, Cancer, Cellular Markers, Growth and Development, Hypoxia, mTOR Pathway, Neuronal Cell Markers, Phospho Specific, Protein Kinase, Serine Kinases, Signal Transduction, Tyrosine Kinases | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
207 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title