Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKT1, p Ser473 Antibody (104A282) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 11 publications

Supplier:  Novus Biologicals NB10056749SS

 View more versions of this product

Catalog No. NB10056749T


Only null left
Add to Cart

Description

Description

AKT1 Monoclonal specifically detects AKT1 in Human, Mouse, Rat, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

AKT1 (p Ser473)
Monoclonal
1 mg/mL
Western Blot, Immunohistochemistry 1:200 - 1:250, Immunohistochemistry-Paraffin 1:200 - 1:250, Immunohistochemistry-Frozen reported by customer review
AKT, AKT serine/threonine kinase 1, AKT1m, CWS6, EC 2.7.11, EC 2.7.11.1, PKB, PKB alpha, PKBMGC99656, PRKBA, Protein kinase B, Protein Kinase B Alpha, Proto-oncogene c-Akt, RAC, rac protein kinase alpha, RAC-ALPHA, RAC-alpha serine/threonine-protein kinas
Mouse
55.7 kDa
0.025 mg
Angiogenesis, Apoptosis, Autophagy, Cancer, Cellular Markers, Growth and Development, Hypoxia, mTOR Pathway, Neuronal Cell Markers, Phospho Specific, Protein Kinase, Serine Kinases, Signal Transduction, Tyrosine Kinases
207
Human, Mouse, Rat, Rabbit
Purified
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
104A282
Unconjugated
P31749
AKT1
This AKT1 phospho Ser473 antibody was raised against a synthetic peptide containing phosphorylated serines at amino acid residues 473 of human AKT1.
Protein G purified
RUO
Primary
Clone 104A282 detects specifically the Ser473 phosphorylated form of AKT1.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only