Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AKT1, p Ser473 Antibody (104A282), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10056749H
Description
AKT1 Monoclonal specifically detects AKT1 in Human, Mouse, Rat, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
AKT1 (p Ser473) | |
Monoclonal | |
HRP | |
P31749 | |
AKT1 | |
This AKT1 phospho Ser473 monoclonal antibody was raised against a synthetic peptide containing phosphorylated serines at amino acid residues 473 of human AKT1. | |
Protein G purified | |
RUO | |
207 | |
Human, Mouse, Rat, Rabbit | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
104A282 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
AKT, AKT serine/threonine kinase 1, AKT1m, CWS6, EC 2.7.11, EC 2.7.11.1, PKB, PKB alpha, PKBMGC99656, PRKBA, Protein kinase B, Protein Kinase B Alpha, Proto-oncogene c-Akt, RAC, rac protein kinase alpha, RAC-ALPHA, RAC-alpha serine/threonine-protein kinas | |
Mouse | |
55.7 kDa | |
0.1 mL | |
Primary | |
Clone 104A282 detects specifically the Ser473 phosphorylated form of AKT1. | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction