Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKT1 Rabbit anti-Human, Mouse, APC, Clone: C7, Abnova™

Catalog No. 89335616 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-335-616 10 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-335-616 Supplier Abnova Corporation Supplier No. MAB18782
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

AKT1 Monoclonal antibody specifically detects AKT1 in Human, Mouse samples. It is validated for Flow Cyt.

Specifications

Antigen AKT1
Applications Flow Cytometry
Classification Monoclonal
Clone C7
Conjugate APC
Description v-akt murine thymoma viral oncogene homolog 1
Gene AKT1
Gene Alias AKT, MGC99656, PKB, PKB-ALPHA, PRKBA, RAC, RAC-ALPHA
Host Species Rabbit
Immunogen A synthetic phosphopeptide corresponding to residues surrounding S473 of human AKT1
Quantity 10 Reactions
Primary or Secondary Primary
Gene ID (Entrez) 207
Target Species Human, Mouse
Content And Storage Store at 4°C
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.