Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AKT1, Rabbit anti-Human, PE, Clone: C7, Abnova™
Rabbit Monoclonal Antibody
Supplier: Abnova Corporation MAB18786
Specifications
| AKT1 | |
| Monoclonal | |
| PE | |
| In PBS, pH 7.4 (0.2% BSA, 0.09% sodium azide). | |
| AKT/MGC99656/PKB/PKB-ALPHA/PRKBA/RAC/RAC-ALPHA | |
| Rabbit | |
| 10 Reactions | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 κ |
| Flow Cytometry | |
| C7 | |
| Flow Cytometry (5 uL/106 cells) The optimal working dilution should be determined by the end user. | |
| v-akt murine thymoma viral oncogene homolog 1 | |
| AKT1 | |
| A synthetic phosphopeptide corresponding to residues surrounding S473 of human AKT1. | |
| RUO | |
| 207 | |
| Store at 4°C. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction