Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AKT1 Rabbit anti-Human, Unconjugated, Clone: RM336, Abnova™

Catalog No. 89339184 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-184 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-184 Supplier Abnova Corporation Supplier No. MAB21967
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

AKT1 Monoclonal antibody specifically detects AKT1 in Human samples. It is validated for ELISA, IHC-P, WB-Tr.

Specifications

Antigen AKT1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone RM336
Conjugate Unconjugated
Description v-akt murine thymoma viral oncogene homolog 1
Gene AKT1
Gene Alias AKT, MGC99656, PKB, PKB-ALPHA, PRKBA, RAC, RAC-ALPHA
Host Species Rabbit
Immunogen A peptide corresponding to AKT1 (E17K mutant)
Quantity 50 μg
Primary or Secondary Primary
Gene ID (Entrez) 207
Target Species Human
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.