Learn More
Invitrogen™ AKT1 Recombinant Rabbit Monoclonal Antibody (RM252)

Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA562116

Description
This antibody reacts to human Akt1. This antibody may also react to bovine, mouse or rat Akt1, as predicted by immunogen homology. Predicted Reactivity: Mouse, Rat.
AKT1 (PKB alpha) is a serine/threonine kinase that regulates cell survival. The activated enzyme inhibits apoptosis and stimulates cell cycle progression by phosphorylating numerous targets in various cell types, including cancer cells. This protein kinase is activated by insulin, PI3K, IGF1 and various other growth and survival factors. Akt promotes cell survival by inhibiting apoptosis through phosphorylation and inactivation of several targets, including forkhead transcription factors, and caspase-9. The AKT pathway is a major target for cancer drug discovery.
Specifications
AKT1 | |
Recombinant Monoclonal | |
Conc. Not Determined | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide | |
P31749 | |
AKT1 | |
A synthetic peptide corresponding to the C-terminus of human Akt1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
RM252 | |
Unconjugated | |
AKT1 | |
Akt; Akt kinase; AKT serine/threonine kinase 1; Akt/PKB; akt1; AKT1 kinase; AKT1m; Akt1m protein; Akt1-PA; Akt1-PB; Akt1-PC; Akt1-PD; Akt1-PE; CAKT; C-AKT; CG4006; CG4006-PA; CG4006-PB; CG4006-PC; CG4006-PD; CG4006-PE; CWS6; dAkt; D-Akt; dAkt kinase; dAKT/dPKB; dAkt/PKB; DAKT1; DAKT1/PKB; Dmel\CG4006; Dmel_CG4006; dPKB; DRAC-PK; DRAC-PK66; DRAC-PK85; l(3)04226; l(3)89Bq; murine thymoma viral (v-akt) oncogene homolog 1; pAkt; P-AKT; PKB; PKB alpha; PKB beta; PKB gamma; PKB/AKT; PKB/dAKT; PKBalpha; PKB-ALPHA; PKBG; PRKBA; protein kinase B; Protein kinase B alpha; protein kinase B-alpha; proto-oncogene c-Akt; RAC; rac protein kinase alpha; RAC protein kinase alpha RAC-PK alpha; RAC serine/threonine protein kinase; RAC serine/threonine-protein kinase; RAC-ALPHA; RAC-alpha serine/threonine-protein kinase; RacPK; RAC-PK-alpha; RAC-PK-beta; RAC-PK-gamma; related to A and C kinases; related to PKA to PKC protein kinases; related to the A and C kinases; SD10374p; serine/threonine protein kinase; serine-threonine protein kinase; Thymoma viral proto-oncogene; thymoma viral proto-oncogene 1; v-akt murine thymoma viral oncogene homolog 1; v-akt murine thymoma viral oncogene-like protein 1 | |
Rabbit | |
Protein A | |
RUO | |
207 | |
-20°C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.