Learn More
Invitrogen™ AKT2 Monoclonal Antibody (1B6)

Description
MA5-15593 targets AKT2 in indirect ELISA, IF, IHC, and WB applications and shows reactivity with Human, Non-human primate, and Rat samples. The MA5-15593 immunogen is purified recombinant fragment of human AKT2 expressed in E. Coli. MA5-15593 detects AKT2 which has a predicted molecular weight of approximately 60kDa.
Specifications
Specifications
| Antigen | AKT2 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 1B6 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | ascites with 0.03% sodium azide |
| Gene | Akt2 |
| Gene Accession No. | P31751, P47197 |
| Gene Alias | 2410016A19Rik; AKT serine/threonine kinase 2; Akt2; AKT2 kinase; AW554154; HIHGHH; murine thymoma viral (v-akt) homolog-2; murine thymoma viral (v-akt) oncogene homolog 2; PKB; PKB beta; PKBB; PKBBETA; PRKBB; protein kinase Akt-2; Protein kinase B beta; protein kinase B, beta; putative v-akt murine thymoma viral oncoprotein 2; rac protein kinase beta; RAC protein kinase beta RAC-PK beta; RAC-BETA; RAC-beta serine/threonine protein kinase; RAC-beta serine/threonine-protein kinase; RAC-PK-beta; thymoma viral proto-oncogene 2; v-akt murine thymoma viral oncogene homolog 2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.