Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat Akt2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB2315SP
This item is not returnable.
View return policy
Description
AKT2 Monoclonal specifically detects AKT2 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
| AKT2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| P31751 | |
| AKT2 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Akt2 Asn2-Glu481 Accession # P31751 | |
| 25 μg | |
| Primary | |
| Detects human, mouse, and rat Akt2. Detects human Akt2 in direct ELISAs and endogenous Akt2 in Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Akt1 or rhAkt3 is observed. | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| 302514 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 2.7.11, EC 2.7.11.1, Murine thymoma viral (v-akt) homolog-2, PKB beta, PKBB, PKBBETA, PRKBB, Protein kinase Akt-2, Protein kinase B beta, rac protein kinase beta, RAC-BETA, RAC-beta serine/threonine-protein kinase, RAC-PK-beta, v-akt murine thymoma viral oncogene homolog 2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 208 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction