Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AKT2, Rabbit anti-Human, Clone: D16-H, Abnova™
Rabbit Monoclonal Antibody
Supplier: Abnova Corporation MAB15923
Specifications
AKT2 | |
Monoclonal | |
Unconjugated | |
In 20 mM Tris-HCl buffer, pH 8.0 (10 mg/mL BSA, 0.05% Sodium Azide). | |
PKBB/PKBBETA/PRKBB/RAC-BETA | |
Rabbit | |
50 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot | |
D16-H | |
ELISA (1:100000-1:200000) Immunocytochemistry Immunoprecipitation Western Blot (1:500) The optimal working dilution should be determined by the end user. | |
v-akt murine thymoma viral oncogene homolog 2 | |
AKT2 | |
A synthetic peptide corresponding to amino acids 455-468 at C-terminus of human AKT2. | |
RUO | |
208 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction