Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                AKT2, p Ser474 Antibody, Novus Biologicals™
                                
                                
                                
                                
                            
                            
                            
                                
                                    
Rabbit Polyclonal Antibody
$268.00 - $426.00
Specifications
| Antigen | AKT2 (p Ser474) | 
|---|---|
| Concentration | 1.0 mg/mL | 
| Dilution | Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100 | 
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | 
| Classification | Polyclonal | 
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
                
                
                    
                        NB017819
                    
                    
                        
                    
                    
                        ![]()  | 
            
                
                    
                        Novus Biologicals
                         NB100817800.025ML  | 
                
            
            
            
            
            
                
                    0.025 mL | 
                    
                        
                        
                            
                             
                                        
                                            
                                                
                                                
                                                
                                            
                                        
                                        
                                            
                                            
                                                
                                                
                                            
                                        
                                        
                                            
                                                
                                                Each for $268.00
                                                 
                                
                             | 
                
                    
                        
                             | 
                    
                
                
                    |||||
                
                
                    
                        NB10081780
                    
                    
                        
                    
                    
                        ![]()  | 
            
                
                    
                        Novus Biologicals
                         NB10081780  | 
                
            
            
            
            
            
                
                    0.05 mL | 
                    
                        
                        
                            
                             
                                        
                                            
                                                
                                                
                                                
                                            
                                        
                                        
                                            
                                            
                                                
                                                
                                            
                                        
                                        
                                            
                                                
                                                Each for $426.00
                                                 
                                
                             | 
                
                    
                        
                             | 
                    
                
                
                    |||||
Description
AKT2 Polyclonal specifically detects AKT2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| AKT2 (p Ser474) | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100 | |
| Polyclonal | |
| Rabbit | |
| Cancer, mTOR Pathway, Phospho Specific | |
| P31751 | |
| 208 | |
| The antiserum was produced against synthesized phosphopeptide derived from human Akt2 around the phosphorylation site of serine 474 (Q-F-SP-Y-S). | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | 
| 1.0 mg/mL | |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| RUO | |
| Rat, Mouse, Human | |
| EC 2.7.11, EC 2.7.11.1, Murine thymoma viral (v-akt) homolog-2, PKB beta, PKBB, PKBBETA, PRKBB, Protein kinase Akt-2, Protein kinase B beta, rac protein kinase beta, RAC-BETA, RAC-beta serine/threonine-protein kinase, RAC-PK-beta, v-akt murine thymoma viral oncogene homolog 2 | |
| AKT2 | |
| IgG | |
| Affinity Purified | |
| 60 kDa | 
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
            
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title