Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AKT3 Antibody (199822) [Alexa Fluor™ 532], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB14631X
Description
AKT3 Monoclonal antibody specifically detects AKT3 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOFSpecifications
AKT3 | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry (Intracellular Staining), CyTOF | |
199822 | |
Intracellular Staining by Flow Cytometry, CyTOF-ready | |
Akt-3, EC 2.7.11, EC 2.7.11.1, PKB gamma, PKBGDKFZp434N0250, PRKBG, v-akt murine thymoma viral oncogene homolog 3 (protein kinase B, gamma) | |
E. coli-derived recombinant human Akt3, Arg23-Ala101, Accession # NP_005456 | |
0.1 mL | |
Cancer, Hypoxia, Protein Kinase, Signal Transduction | |
10000 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction