Learn More
Promega AKT3 Kinase Enzyme System, 10microg
Catalog No. PRV4010 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV4010 10 μg
1 options
Catalog No. PRV4010 Supplier Promega Supplier No. V4010
Recombinant full-length human AKT3 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. AKT3 or Protein Kinase B gamma (PKBgamma) is a serine/threonine kinase that is a member of the AKT family.
  • Easily Screen and Profile AKT3 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C