Learn More
Promega AKT3 Kinase Enzyme System, 1mg
Catalog No. PRV6271 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6271 1 mg
1 options
Catalog No. PRV6271 Supplier Promega Supplier No. V6271
Add to Cart
Add to Cart
Request a Quote
Recombinant full-length human AKT3 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. AKT3 or Protein Kinase B gamma (PKBgamma) is a serine/threonine kinase that is a member of the AKT family.
  • Easily Screen and Profile AKT3 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C