Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AKTIP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559734
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559734 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559734 Supplier Invitrogen™ Supplier No. PA559734
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WFGVIFIRHG LYQDGVFKFT VYIPDNYPDG DCPRLVFDIP VFHPLVDPTS GELDVKRAFA KWRRNHNHIW QVLMYARRVF YKIDTASP Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

Component of the FTS/Hook/FHIP complex (FHF complex). The FHF complex may function to promote vesicle trafficking and/or fusion via the homotypic vesicular protein sorting complex (the HOPS complex). Regulates apoptosis by enhancing phosphorylation and activation of AKT1. Increases release of TNFSF6 via the AKT1/GSK3B/NFATC1 signaling cascade.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AKTIP
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AKTIP
Gene Accession No. Q9H8T0
Gene Alias AKT interacting protein; AKT-interacting protein; AKTIP; AL023020; Fif; Ft; FT1; FTS; fused toes homolog; Fused toes protein; Fused toes protein homolog; thymoma viral proto-oncogene 1 interacting protein
Gene Symbols AKTIP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human AKTIP. Recombinant protein control fragment (Product #RP-98740).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64400
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.