Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ALAD Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$382.00 - $646.00
Specifications
Antigen | ALAD |
---|---|
Dilution | Western Blot 0.4 ug/ml, Simple Western 1:200, Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
ALAD Polyclonal specifically detects ALAD in Human, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ALAD | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Rat | |
ALADHEC 4.2.1.24, aminolevulinate dehydratase, aminolevulinate, delta-, dehydratase, delta-aminolevulinic acid dehydratase, PBGSMGC5057, Porphobilinogen synthase | |
ALAD | |
IgG | |
Affinity Purified | |
Specificity of human ALAD antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot 0.4 ug/ml, Simple Western 1:200, Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Polyclonal | |
Rabbit | |
Proteases & Other Enzymes | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
210 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:GCQVVAPSDMMDGRVEAIKEALMAHGLGNRVSVMSYSAKFASCFYGPFRDAAKSSPAFGDRRCYQLPPGARGLALRAVDRD | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title