Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ALAD Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554310
Description
Immunogen sequence: GCQVVAPSDM MDGRVEAIKE ALMAHGLGNR VSVMSYSAKF ASCFYGPFRD AAKSSPAFGD RRCYQLPPGA RGLALRAVDR D Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.
The ALAD enzyme is composed of 8 identical subunits and catalyzes the condensation of 2 molecules of delta-aminolevulinate to form porphobilinogen (a precursor of heme, cytochromes and other hemoproteins). ALAD catalyzes the second step in the porphyrin and heme biosynthetic pathway; zinc is essential for enzymatic activity. ALAD enzymatic activity is inhibited by lead and a defect in the ALAD structural gene can cause increased sensitivity to lead poisoning and acute hepatic porphyria.The ALAD enzyme is composed of 8 identical subunits and catalyzes the condensation of 2 molecules of delta-aminolevulinate to form porphobilinogen (a precursor of heme, cytochromes and other hemoproteins). ALAD catalyzes the second step in the porphyrin and heme biosynthetic pathway; zinc is essential for enzymatic activity. ALAD enzymatic activity is inhibited by lead and a defect in the ALAD structural gene can cause increased sensitivity to lead poisoning and acute hepatic porphyria. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
ALAD | |
Polyclonal | |
Unconjugated | |
ALAD | |
Alad; ALADH; ALADR; aminolevulinate dehydratase; aminolevulinate, delta-, dehydratase; aminolevulinatedelta-dehydratase; delta - aminolevulinic acid dehydratase; delta-aminolevulinic acid dehydratase; Lv; PBGS; Porphobilinogen synthase; testicular tissue protein Li 95 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
210 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P13716 | |
ALAD | |
Recombinant protein corresponding to Human ALAD. Recombinant protein control fragment (Product #RP-93005). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction