Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALAD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554564
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554564 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554564 Supplier Invitrogen™ Supplier No. PA554564
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VLIFGVPSRV PKDERGSAAD SEESPAIEAI HLLRKTFPNL LVACDVCLCP YTSHGHCGLL SENGAFRAEE SRQRLAEVAL AYA Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.

The ALAD enzyme is composed of 8 identical subunits and catalyzes the condensation of 2 molecules of delta-aminolevulinate to form porphobilinogen (a precursor of heme, cytochromes and other hemoproteins). ALAD catalyzes the second step in the porphyrin and heme biosynthetic pathway; zinc is essential for enzymatic activity. ALAD enzymatic activity is inhibited by lead and a defect in the ALAD structural gene can cause increased sensitivity to lead poisoning and acute hepatic porphyria.The ALAD enzyme is composed of 8 identical subunits and catalyzes the condensation of 2 molecules of delta-aminolevulinate to form porphobilinogen (a precursor of heme, cytochromes and other hemoproteins). ALAD catalyzes the second step in the porphyrin and heme biosynthetic pathway; zinc is essential for enzymatic activity. ALAD enzymatic activity is inhibited by lead and a defect in the ALAD structural gene can cause increased sensitivity to lead poisoning and acute hepatic porphyria. Alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALAD
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ALAD
Gene Accession No. P13716
Gene Alias Alad; ALADH; ALADR; aminolevulinate dehydratase; aminolevulinate, delta-, dehydratase; aminolevulinatedelta-dehydratase; delta - aminolevulinic acid dehydratase; delta-aminolevulinic acid dehydratase; Lv; PBGS; Porphobilinogen synthase; testicular tissue protein Li 95
Gene Symbols ALAD
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ALAD. Recombinant protein control fragment (Product #RP-93004).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 210
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.