Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Aladin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559060
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559060 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559060 Supplier Invitrogen™ Supplier No. PA559060
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QLITFHPSFN KGALLSVGWS TGRIAHIPLY FVNAQFPRFS PVLGRAQEPP AGGGGSIHDL PLFTETSPTS APWDPLPGPP PVLPHS Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 86%.

The protein encoded by this gene is a member of the WD-repeat family of regulatory proteins and may be involved in normal development of the peripheral and central nervous system. The encoded protein is part of the nuclear pore complex and is anchored there by NDC1. Defects in this gene are a cause of achalasia-addisonianism-alacrima syndrome (AAAS), also called triple-A syndrome or Allgrove syndrome. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aladin
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AAAS
Gene Accession No. Q9NRG9
Gene Alias AAA; Aaas; AAASb; achalasia, adrenocortical insufficiency, alacrimia; achalasia, adrenocortical insufficiency, alacrimia (Allgrove, triple-A); ADRACALA; Adracalin; ALADIN; aladin WD repeat nucleoporin; Allgrove, triple-A; D030041N15Rik; GL003
Gene Symbols AAAS
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Aladin. Recombinant protein control fragment (Product #RP-97679).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8086
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.