Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Aladin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109350
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109350 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109350 Supplier Invitrogen™ Supplier No. PA5109350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GGGVTNLLWS PDGSKILATT PSAVFRVWEA QMWTCERWPT LSGRCQTGCW SPDGSRLLFT VLGEPLIYSL SFPERCGEGK GCVGGAKSAT IVADLSETTI QTPDGEERLG GEAHSMVWDP SGERLAVLMK GKPRVQDGKP VILLFRTRNS PVFELLPCGI IQGEPGAQPQ LITFHPSFNK GALLSVGWST GRIAHIPLYF VNAQFPRFSP VLGRAQEPPA GGGGSIHDLP LFTETSPTSA PWDPLPGPPP VLPHSPHSHL.

The protein encoded by this gene is a member of the WD-repeat family of regulatory proteins and may be involved in normal development of the peripheral and central nervous system. The encoded protein is part of the nuclear pore complex and is anchored there by NDC1. Defects in this gene are a cause of achalasia-addisonianism-alacrima syndrome (AAAS), also called triple-A syndrome or Allgrove syndrome. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aladin
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene AAAS
Gene Accession No. Q9NRG9
Gene Alias AAA; Aaas; AAASb; achalasia, adrenocortical insufficiency, alacrimia; achalasia, adrenocortical insufficiency, alacrimia (Allgrove, triple-A); ADRACALA; Adracalin; ALADIN; aladin WD repeat nucleoporin; Allgrove, triple-A; D030041N15Rik; GL003
Gene Symbols AAAS
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 287-546 of human AAAS (NP_056480.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8086
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.