Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Aladin Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA550308

Catalog No. PA550308


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total Aladin protein.

The protein encoded by this gene is a member of the WD-repeat family of regulatory proteins and may be involved in normal development of the peripheral and central nervous system. The encoded protein is part of the nuclear pore complex and is anchored there by NDC1. Defects in this gene are a cause of achalasia-addisonianism-alacrima syndrome (AAAS), also called triple-A syndrome or Allgrove syndrome. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Aladin
Polyclonal
Unconjugated
AAAS
AAA; Aaas; AAASb; achalasia, adrenocortical insufficiency, alacrimia; achalasia, adrenocortical insufficiency, alacrimia (Allgrove, triple-A); ADRACALA; Adracalin; ALADIN; aladin WD repeat nucleoporin; Allgrove, triple-A; D030041N15Rik; GL003
Rabbit
Antigen affinity chromatography
RUO
8086
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q9NRG9
AAAS
Fusion protein corresponding to residues near the C terminal of human achalasia, adrenocortical insufficiency, alacrimia.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.