Learn More
Pyramex™ Alair™ Safety Eyewear
A stylish look that is 100% polycarbonate. These economical glasses have a hard coated lens-unlike most at this price
$79.37 - $92.51
Specifications
Certifications/Compliance ANSI Z87.1
Lens Material Polycarbonate
View More Specs
Products 2
Catalog Number Mfr. No. Lens Tint Price Quantity & Availability  
Catalog Number Mfr. No. Lens Tint Price Quantity & Availability  
19-155-550
View Documents
Pyramex™
S3210ST
Clear
Case of 12 for $79.37
Only null left
Add to Cart
19-155-553
View Documents
Pyramex™
S3270S
Silver, Mirror
Case of 12 for $92.51
Only null left
Add to Cart
Description
Description
  • Economically priced to fit any budget
  • Incredibly light, resilient & durable protective eyewear for all day wear
  • Closefitting unitary front with dual 9.5 base curvature offers wide and unobstructed vision
  • Fashionable design resembles sunglasses, suitable for both men and women
  • Scratch-resistant, hard-coated polycarbonate lens filter out more than 99.9% of harmful UV rays
  • Specifications
    Specifications
    ANSI Z87.1
    Polycarbonate
    Videos
    SDS
    Documents
    Documents
    Product Certifications
    Product Content Correction
    Your input is important to us. Please complete this form to provide feedback related to the content on this product.
    Product Title
    Your feedback has been submitted: Thank you for helping us improve our website.