Learn More
Thermo Scientific™ alamarBlue™ Dye
Allows three assays to be performed with just one dye, including cell growth determination, in vitro cytotoxicity and cytokine assays.
$744.00 - $973.00
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
10-230-103
|
Thermo Scientific™
Y00010 |
10 mL |
Each for $744.00
|
|
|||||
10-230-102
|
Thermo Scientific™
Y00100 |
100 mL |
Each for $973.00
|
|
|||||
Description
Thermo Scientific™ alamarBlue provides unmatched sensitivity and versatility, allowing three assays to be performed with just one dye, including cell growth determination, in vitro cytotoxicity and cytokine assays.
alamarBlue, a resazurin-based product with extended incubation time, is stable for up to 5 days.Specifications
alamarBlue indicator dye |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.