Learn More
Thermo Scientific Chemicals β-Alanine, 98%
Description
Effective catalyst for the Knoevenagel condensation. Beta Alanine is used as nutrition supplements in food production. Increase muscular strength & power output, Increases Muscle Mass, increase Anaerobic Endurance.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Effective catalyst for the Knoevenagel condensation. Beta Alanine is used as nutrition supplements in food production. Increase muscular strength & power output, Increases Muscle Mass, increase Anaerobic Endurance.
Solubility
Soluble in water(550g/L). Slightly soluble in alcohol. Insoluble in ether and acetone.
Notes
Slightly hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Specifications
Specifications
Melting Point | ∼205°C (decomposition) |
Assay Percent Range | 98% |
Linear Formula | H2NCH2CH2CO2H |
Beilstein | 906793 |
Merck Index | 14,205 |
Solubility Information | Soluble in water(550g/L). Slightly soluble in alcohol. Insoluble in ether and acetone. |
Density | 1.437 |
Quantity | 1000 g |
Odor | Odorless |
Formula Weight | 89.09 |
Show More |
Safety and Handling
EINECSNumber | 203-536-5 |
RTECSNumber | UA2369200 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.