Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ alanyl-tRNA Synthetase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559335
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559335 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559335 Supplier Invitrogen™ Supplier No. PA559335
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DRASKADVQK RVLEKTKQFI DSNPNQPLVI LEMESGASAK ALNEALKLFK MHSPQTSAML FTVDNEAGKI TCLCQVPQNA ANRGLKASEW VQQVSGLMDG KGGGKD Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

The human alanyl-tRNA synthetase (AARS) belongs to a family of tRNA synthases, of the class II enzymes. Class II tRNA synthases evolved early in evolution and are highly conserved. This is reflected by the fact that 498 of the 968-residue polypeptide human AARS shares 41% identity witht the E. coli protein. tRNA synthases are the enzymes that interpret the RNA code and attach specific aminoacids to the tRNAs that contain the cognate trinucleotide anticodons. They consist of a catalytic domain which interacts with the amino acid acceptor-T psi C helix of the tRNA, and a second domain which interacts with the rest of the tRNA structure.
TRUSTED_SUSTAINABILITY

Specifications

Antigen alanyl-tRNA Synthetase
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AARS1
Gene Accession No. P49588, P50475, Q8BGQ7
Gene Alias Aars; AI316495; alanine tRNA ligase 1, cytoplasmic; alanine--tRNA ligase, cytoplasmic; alanine--tRNA ligase, cytoplasmic {ECO:0000255; Alanyl-tRNA synthetase; alanyl-tRNA synthetase {ECO:0000255; alanyl-tRNA synthetase, cytoplasmic; alaRS; alaRS {ECO:0000255; C76919; CMT2N; EIEE29; HAMAP-Rule:MF_03133}; Protein sticky; Renal carcinoma antigen NY-REN-42; sti
Gene Symbols AARS1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human alanyl-tRNA Synthetase. Recombinant protein control fragment (Product #RP-98616).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16, 234734, 292023
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.