Learn More
BINDER™ Alarm Output
SDP
Catalog No. p-200104331 Shop All BINDER Inc. Products
Click to view available options
For Use With (Equipment):
KB 65/130/260/470/720/1060
KB PRO Series
KBF PRO Series
KBF Series
Alarm output, zero-voltage, accessible via 3-pin clamping sleeve
  • Alarm output
  • Zero-voltage
  • Accessible via 3-pin clamping sleeve (max. 24V/2.5A)
  • Acoustic signal that can be switched off
Specifications
For Use With (Equipment) KB 65/130/260/470/720/1060
Material Aluminum
Type Analog Output
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.