Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alas1 Rabbit anti-Human, Clone: 2K1G3, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | Alas1 |
---|---|
Clone | 2K1G3 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Alas1 Monoclonal antibody specifically detects Alas1 in Human samples. It is validated for Western BlotSpecifications
Alas1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cancer, Cardiovascular Biology, Endocrinology, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
211 | |
IgG | |
Affinity purified |
2K1G3 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
5-aminolevulinic acid synthase 1, ALAS, ALAS3, ALASH, ALAS-H, aminolevulinate, delta-, synthase 1, Delta-ALA synthase 1,5-aminolevulinate synthase, nonspecific, mitochondrial, Delta-aminolevulinate synthase 1, EC 2.3.1, EC 2.3.1.37, MIG4, migration-inducing protein 4 | |
A synthetic peptide corresponding to a sequence within amino acids 541-640 of human Alas1 (P13196). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title